Learn More
Motic™ Moticam 1080X Camera
The Moticam 1080X Camera that designed for ease of use and the ability to output fast live images at 1920 x 1080 pixels in full HD. Simply plug in the camera through the HDMI port and display images with the use of the onboard software and a USB mouse through an HDMI monitor.
Supplier: Motic™ 1100600101482
This item is not returnable.
View return policy
Description
- C-mount for lens
- MotiConnect software is available for free download (with free updates included)
- 8 MP resolution on SD card (3840 x 2160 px)
Specifications
Microscope Camera | |
8 MP | |
1/1.9 in. |
12 V Power Adapter, CS Ring, Motic™ 4-dot Calibration Slide, Dust Cap, Manual, 12 mm Lens, 30 mm to 38 mm Adapter, Macro Tube, Macro Calibration Card | |
IMX185 |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov