Learn More
Fisherbrand™ Mortar and Pestle Deep Form
Mortar and Pestle Deep Form
 View more versions of this product
Catalog No. FB970R
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative. View substitute products »
Description
Description
  • Low permeability
  • High strength
  • Hard
  • High resistance to chemical attacks
  • Translucent or glazed body
Specifications
Specifications
120mm
Agate
Mortar and Pestle
Mortar and Pestle Set
For grinding or crushing