Learn More
Invitrogen™ Live Cell Imaging Solution
Physiological medium developed for live cell imaging applications that has broad utility in cell-based research, including dye loading and wash steps
Supplier: Invitrogen™ A14291DJ
Description
Live cell imaging can be challenging. Not only must staining be optimized for particular assay readout, spectral compatibility, and signal-to-noise level, but live cell imaging of dynamic processes and cell physiology requires live observation over time. The choice of medium is particularly important for time-lapse imaging and experiments where cells are exposed to ambient conditions for longer periods. For reliable results, it is essential that cells be healthy and maintained as closely as possible to physiological temperature, pH, oxygen tension, and other conditions. If cells appear unhealthy they should not be used for imaging.
- Optically clear solution buffered with HEPES at pH 7.4 that keeps cells healthy for up to 4 hours at ambient atmosphere and temperature
- Suitable solution for live cell fluorescence imaging applications
- Extended buffering capacity compared to media for improved cell viability
- Developed specifically for live cell imaging applications: provides image clarity, signal-to-noise ratio, and cell viability that are significantly enhanced compared to standard media
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Sample Preparation
Order Info
Shipping Condition: Room temperature
Specifications
Live Cell Imaging Solution | |
Physiological Imaging Buffer | |
Sustainable packaging | |
Room Temperature |
Molecular Probes | |
Store at ≤25 °C | |
500 mL |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.