Learn More
Moldex™ Meteors™ PlugStation™ Ear Plug Dispenser
Click to view available options
500 Pairs/Dispenser, 4 Dispensers/Cs.
Description
- Holds Meteors™ disposable ear plugs
- PlugStations™ are sealed to ensure ear plugs stay clean and sanitary
- High visibility to increase compliance
- Dispenses one pair at a time to eliminate wasteful grabbing
- Includes mounting bracket
Specifications
Includes | Mounting Hardware |
ANSI S3.1974 | Yes |
CSA Z94.2 | Class AL |
Size | Large |
For Use With (Application) | Easy Access to Hearing Protection for Workers and Facility Visitors in Reception Areas, Break Rooms, Workstations, and Other Noisy Environments. |
Holds | 500 Pairs |
Pairs per Package | 500 Pairs/Dispenser |
Plug Material | Foam |
Plug Color | Green |
Noise Reduction Rating (NRR) | 33 dB |
Show More |