Learn More
SCS Moisture Sensitive Caution Label (IPC/JEDEC J-STD-020)
Label is used to identify moisture sensitive devices for levels 2, 2a, 3, 4, 5, and 5a as defined by J-STD-020
Supplier:  SCS 113LABEL
Catalog No. 18-688-206
Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative.
Description
Description
  • 4 Inch Width x 4 Inch Height JEDEC (JEP113-B) Moisture Sensitive Caution Label
    Used on Foil Moisture Barrier Bags to identify moisture sensitive devices for levels 2, 2a, 3, 4, 5, and 5a as defined by J-STD-020.
  • Permanent Adhesive
    For one-time use when the label will remain intact and never be removed.
  • 100 per Roll
  • Made in the United States of America
    This item is stocked in Sanford, NC
Specifications
Specifications
Not Autoclavable
White
Non-sterile
Moisture Barrier Bags
100/Roll
10.16 cm
Permanent
Paper
Caution: This bag contains MOISTURE-SENSITIVE DEVICES
Yes
4 x 4 in.
10.16 cm