Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EVAC+CHAIR™ Model 500H Bariatric Evac+Chair
SDPSCI_ED

Two Person Operation Enhanced Evac+Chair is for use on stairs between 28 and 45°

Supplier:  EVAC+CHAIR™ 500H

Catalog No. S72765


Add to Cart

Alert:

Emergency Prepardness, Evacuation


Description

Description

  • Passenger weight capacity:  500lbs
  • Can be used as a single person operation chair for payloads up to 500 lbs
  • Emergency Evacuation Chair
  • Adjustable patient head restraint
  • Friction tracks (Evac+Track technology)
  • Ergonomic adjustable handle
  • Reflective decals
  • Quick release safety belts (Head, Chest, Thigh)
  • Heavy duty wheels
  • Lockable rear wheels
  • Dold away grip handle/footrest
  • Footrest
  • Dual Position Seating
  • Vinyl Storage Cover
  • Mounting Wall Bracket
  • Photo-luminescent Indicator Sign
  • Instructional DVD
  • Full Color User Guide
  • Lifetime Warranty
Specifications

Specifications

20 in.
45 in.
9 in.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

The World’s No.1 Stairway Evacuation Chair Since 1982