Learn More
Ludlum Measurements Inc Model 2241-2 Digital Scaler-Ratemeter
Supplier: Ludlum Measurements Inc 482731
This instrument is a general purpose, digital ratemeter with a built-in scaler that provides timed counts over a user specified period. This system will store detector setup information for up to two detectors so personnel can easily change detectors in the field. The LCD presents readings in a four-digit moving decimal format together with one of a wide range of available units of measure and is accompanied by several operational and alert icons. The LCD backlight can be activated and remains lit for a user-adjustable interval. This instrument features adjustable alert and alarm setpoints for both the Ratemeter and Scaler modes. Front-panel controls include a rotary switch for selecting between Off, Ratemeter Mode, and Scaler Mode; switches for audio on/off and fast/variable response time; and push-buttons for activating the LCD backlight and for either resetting the counts or acknowledging alarms.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.