Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LifeSecure™ MobileAid™ Grab-N-Go Trauma First Aid Field Kit



157-piece Grab-and-Go Trauma First Aid Kit with essential trauma first aid supplies organized in four expandable compartments.
Supplier: LifeSecure™ 37320
Description
- 157-piece
- Contains essential trauma first aid supplies
- Shoulder bag with four expandable compartments
- Compact storage to be ready when needed
- Patterned after military-style medic field trauma bags with quick release buckle
- Rugged 1000D construction
Specifications
First Aid Kit | |
17.78 cm | |
1 Ea. | |
22.86 cm |
First Aid | |
Red | |
7 x 9 x 4 in. | |
10.16 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title