Learn More
Seca Corp 217 Stable stadiometer for mobile height measurement
Supplier: Seca Corp 2171821009
The mobile stadiometer seca 217 boasts unmatched stability, a cleverly designed assembly system and particularly high-quality materials. All this lends the stadiometer an exceptionally high degree of stability – almost the same as a permanent installation. In addition, the adjustable spacer ensures a secure hold without any fittings. Suitable for medical practices and hospitals, the stadiometer is also just right for mobile use, such as examinations of children at school or patients at home. • Parts fit together perfectly for easy transport • Wall spacer for additional stability • Robust platform for secure stance • Includes heel positioner for optimal three-point height measurement • Adapter element seca 437 for combination with a flat scale • Measuring range: 8 inches – 6 ft 9 inches / 20 – 205 cm • Graduation: 1/8 in / 1 mm • Optional: adapter element seca 437 (for combination with flat scale seca 869, seca 874 or seca 876), carrying case seca 414 or seca 412
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.