Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MK 0343, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Subtype-selective GABAA partial agonist (Ki values are 0.21, 0.22, 0.23 and 0.40 for α3, α1, α5 and α2 respectively); occupies the benzodiazepine site of GABAA receptors. Displays greater agonist efficacy at α3 compared to α1. Exhibits anxiolytic and non-sedating properties in rodent models. Brain penetrant.
Specifications
Quantity | 50 mg |
Formula Weight | 397.38 |
Percent Purity | >99% |
Chemical Name or Material | MK 0343 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction