Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Block, 22 mm, 13 mm and 6 mm dia., EMD
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Supplier: Thermo Scientific™ 88870114/EMD
Description
Specifications
Mixed block, 22mm, 13mm and 6mm dia. | |
3 x 25mm, 12 x 12∽13mm and 6 x 6mm | |
76 x 124 x 39 mm (3 x 4.9 x 1.5 in.) |
Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models) | |
Block |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction