Learn More
MSC Mitutoyo 293-130-10 0mm to 25.4mm 0.98" Throat Electronic Outside Micrometer 0.00010mm Resolution, 0.000020" Accuracy, Ratchet-Friction Thimble
Supplier:  MSC 77571875
Encompass
Mitutoyo 293-130-10 0mm to 25.4mm 0.98" Throat Electronic Outside Micrometer 0.00010mm Resolution, 0.000020" Accuracy, Ratchet-Friction Thimble
Catalog No. NC1112073