Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mitchell Plastics Dispensing Bins
Conveniently stores and dispenses many small items
Specifications
Type | Dispensing Bin |
---|---|
Color | Clear |
Shape | Rectangular |
Mounting Type | Wall Mount |
For Use With (Application) | Storage |
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
06-666-82
|
Mitchell Plastics
ML 2550 |
Mini dispensing bin | Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. | N/A | |||||
05-723-452
|
Mitchell Plastics
ML2000 |
Holds 500 disposable pipet tips | Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. | N/A | |||||
Description
- Dust-free storage
- Wall mount or use on bench
- Clear front panels to view contents
- Holds 500 disposable pipet tips
- Removable lid
Specifications
Dispensing Bin | |
Rectangular | |
Storage |
Clear | |
Wall Mount | |
5 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title