Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mirus Bio™ TransIT™-CHO Transfection Kit
TransIT™-CHO Transfection Kit developed for high transfection efficiency and cell viability in CHO cell types. Does not require medium changes and can be performed in serum-containing medium.
$343.00 - $2586.00
Specifications
Content And Storage | Store at -20°C |
---|---|
Cell Type | CHO |
Format | Tube |
Promoter | None (Promoterless) |
Transfection Technique | Chemical Transfection |
Includes: TransIT-CHO reagent and CHO Mojo reagent, both supplied as ready-to-use solutions.
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
MIR-2170
|
Mirus Bio™
MIR2170 |
1 mL 2.22 mg/mL in 80% Ethanol, TransIT™-CHO reagent and 0.70 mL 5 mg/mL in 100% CHO Mojo reagent, both supplied as ready-to-use solution |
Each for $928.00
|
|
|||||
MIR-2174
|
Mirus Bio™
MIR2174 |
0.4 mL 2.22 mg/mL in 80% Ethanol, TransIT™-CHO reagent and 0.28 mL 5 mg/mL in 100% CHO Mojo reagent, both supplied as ready-to-use solution |
Each for $343.00
|
|
|||||
MIR2175
|
Mirus Bio™
MIR 2175 |
5 mL 2.22 mg/mL in 80% Ethanol, TransIT™-CHO reagent and 3.5 mL 5 mg/mL in 100% CHO Mojo reagent, both supplied as ready-to-use solution |
Each for $2,586.00
|
|
|||||
MIR2176
|
Mirus Bio™
MIR2176 |
10 mL 2.22 mg/mL in 80% Ethanol, TransIT™-CHO reagent and 7 mL 5 mg/mL in 100% CHO Mojo reagent, both supplied as ready-to-use solution | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- TransIT-CHO reagent and CHO Mojo reagent, both supplied as ready-to-use solutions.
- Unique Formulation: Each kit contains a base reagent and a booster reagent to optimize transfection performance in CHO cells
- Low Cellular Toxicity: Maintains cell density and reduces experimental bias
- High Efficiency Delivery: Allows 50 to 60% transfection efficiency in CHO-K1 cells
- Suitable for both transient and stable transfection
Specifications
Store at -20°C | |
Tube | |
Chemical Transfection | |
Transfection Reagent | |
Sterile |
CHO | |
None (Promoterless) | |
Specifically optimized to provide exceptional transfection efficiency of plasmid DNA in CHO cells and cell types of associated lineage | |
1 Ea. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title