Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mirus Bio™ Label IT™ RNAi Delivery Controls
Label IT™ RNAi Delivery Controls feature fluorescently labeled RNAi duplexes ideal for In Vitro or In Vivo tracking using direct fluorescent microscopy.
$153.00 - $941.00
Specifications
Format | Liquid |
---|---|
Product Type | Label IT RNAi Delivery Control |
Stability | 6 Months |
Storage Buffer | Dilution Buffer (50 mM Tris, pH 7.5, 100 mM NaCl) |
Content And Storage | -20°C |
Catalog Number | Mfr. No. | Concentration | Label or Dye | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Concentration | Label or Dye | Quantity | Price | Quantity & Availability | |||
MIR-7900
|
Mirus Bio™
MIR 7900 |
0.75 nmol | Cy™3 | 10 μg |
Each for $324.00
|
|
|||
MIR-7903
|
Mirus Bio™
MIR 7903 |
7.5 nmol | Fluorescein | 100 μg |
Each for $767.00
|
|
|||
MIR-7901
|
Mirus Bio™
MIR 7901 |
7.5 nmol | Cy™3 | 100 μg |
Each for $941.00
|
|
|||
MIR-7902
|
Mirus Bio™
MIR 7902 |
0.75 nmol | Fluorescein | 10 μg |
Each for $153.00
|
|
|||
Description
- Sensitive - Easily visualize transfected cells and assess transfection efficiency using fluorescent microscopy
- Inert - Does not target any mammalian genes or cause off-target effects
- Compatible - Suitable for co-delivery experiments with functional siRNA
- Ease of Use - Supplied as a ready to use 10 μM stock solution with a 10X RNAi Dilution Buffer
- Visualization and tracking of transfected RNAi duplexes
Specifications
Liquid | |
6 Months | |
-20°C |
Label IT RNAi Delivery Control | |
Dilution Buffer (50 mM Tris, pH 7.5, 100 mM NaCl) | |
in-vitro or in vivo tracking using direct fluorescent microscopy, co-delivery experiments with functional siRNA |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title