Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Minigrip™ SPECI-ZIP™ Reclosable Colored Biohazard Bags
Designed with info pocket (three wall) and tear line
$112.00 - $120.00
Specifications
Product Type | Reclosable Biohazard Bag |
---|---|
For Use With (Application) | Designed with info pocket, tear line |
Includes | Printed biohazard warning, info pocket, tear line |
Width (English) | 6 in. |
Width (Metric) | 15.24 cm |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
22-310-407
|
Minigrip™
IP69B |
Clear |
Case of 1000 for $112.00
|
|
|||||
22-310-413
|
Minigrip™
IP69BG |
Green |
Case of 1000 for $116.00
|
|
|||||
22-310-412
|
Minigrip™
IP69BY |
Yellow |
Case of 1000 for $117.00
|
|
|||||
22-310-411
|
Minigrip™
IP69BBL |
Blue |
Case of 1000 for $120.00
|
|
|||||
Description
- Colored or clear transparent plastic bags with biohazard logo
- Bilingual in English and Spanish
Order Info
Case of 1000 in packs of 100.
Specifications
Reclosable Biohazard Bag | |
Printed biohazard warning, info pocket, tear line | |
15.24 cm | |
22.86 cm |
Designed with info pocket, tear line | |
6 in. | |
9 in. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title