Learn More
Vector Laboratories Dolichos Biflorus Agglutinin (DBA) Biotinylated 5 mg
Supplier: Vector Laboratories B10355
Dolichos biflorus agglutinin (DBA) binds α-linked N-acetylgalactosamine. Used to establish secretor status in blood group A individuals by hemagglutination inhibition techniques and for blood typing, this lectin has also been used as a general marker of developing renal collecting ducts. This conjugate features a ratio of biotin to lectin protein that provides optimal avidin/streptavidin and lectin binding activity. Supplied essentially free of unconjugated biotins, it is preserved with sodium azide.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.