Learn More
Scigen Microtome Lubrication Kit
Designed for all types of microtomes operating between 80°C to -100°C
Supplier: Scigen 5029
This item is not returnable.
View return policy
Description
- Kit Components: Special low temperature lubricants and grease
- When used regularly, lubricants and grease help reduce gear wear and maintain microtome cutting tolerances
- Lubricants and grease can be used on any gear or moving parts system where extended instrument life is desired
Specifications
Microtome Lubrication Kit |