Learn More
Roboz Surgical MICROSTERILIZING CASE
SDP
Supplier:  Roboz Surgical RS9902A
Encompass
Micro Sterilizing Case With Cushions; 8 X 12.5 X 1.25 in. An Effective Way Of Protecting Delicate Micro Surgical Instruments For Sterilization. Silicone Rubber inserts Protect Instruments. Includes Removable Cover and a Sentry removable silicone mat.
Catalog No. NC0248857