Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synthware™ Microscale Filtering Flask
Flask has a tapered top designed to fit a filter funnel.
Specifications
Color | Clear |
---|---|
Material | Borosilicate Glass |
For Use With (Application) | Organic Synthesis |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
31-502-130
|
Synthware™
F150025 |
25 mL |
Each for $27.25
|
N/A | |||||
31-502-131
|
Synthware™
F150050 |
50 mL |
Each for $27.25
|
N/A | |||||
Specifications
Clear | |
Borosilicate Glass | |
Organic Synthesis |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title