Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Preparation Rack, 96-well, with cover

Thermo Scientific™ 96-well sample preparation rack.

Supplier:  Thermo Scientific™ PRK0963

Catalog No. FERPRK0963


Explore available promotions
Add to Cart

Description

Description

Thermo Scientific™ 96-well sample preparation rack.

For all 0.2mL tubes, strip tubes, 24-well Piko™ PCR Plates and 96-well non-skirted or semi-skirted plates.
  • Blue rack, clear cover
  • Industry-standard footprint fits in microplate centrifuge rotors
  • Clear lid allows for easy stacking and viewing of samples

Includes:

Recommended for:

Sample handling, sample preparation, sample storage

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Microplate Rack
0.2 mL Tubes
Real-Time PCR Systems or Thermal Cyclers
5 racks
Blue/Clear, Clear
Rectangular
5 Racks
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.