Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Greiner Bio-One MICROLON™ 96-well ELISA Single-Break Strip Plates
For use in colorimetric immunoassay. Greiner Bio-One MICROLON™ 96-Well ELISA Single-Break Strip Plates can be broken off separately and number of tests performed can be adjusted to number of samples.
Supplier: Greiner Bio-One 705066
Description
- Free of detectable DNase, RNase, human DNA Non-pyrogenic
- Individual wells can be pipetted without leaving residue
- Flat bottom enables precise optical measurements
- Suitable for transmission measurements
- Specifica
Specifications
Single break | |
Strip Plate | |
Medium Binding | |
Polystyrene | |
Non-sterile | |
C | |
Colorimetric immunoassay | |
Yes |
Medium Binding | |
8 x 12 | |
Yellow, Clear Bottom, Clear Bottom | |
96 | |
Microlon | |
Yes | |
100/Cs. | |
127.76 x 85.48 x 14.5 mm (5.02 x 3.36 x 0.5 in.) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction