Learn More
Microbiologics™ Epower™ Bacillus subtilis subsp. spizizenii ATCC™ 6633™†
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination
Specifications
ATCC® | 6633™* |
---|---|
Biosafety Level | 1 |
Form | Lyophilized Pellet |
Quantity | 10/Pk. |
Catalog Number | Mfr. No. | Quantity | Mean Assay Value | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Mean Assay Value | Price | Quantity & Availability | ||||
12-111-239
|
Microbiologics™
0486E4CRM |
10/Pk. | 1.0-9.9 + 4 CFU/pellet |
Each for $907.50
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
12-111-238
|
Microbiologics™
0486E3CRM |
10/Pk. | 1.0-9.9 + 3 CFU/pellet |
Each for $907.50
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
12-111-240
|
Microbiologics™
0486E6CRM |
10/Pk. | 1.0-9.9 + 6 CFU/pellet |
Each for $907.50
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Epower
- Lyophilized microorganism preparations
- Biosafety Level: 1
- Pellets can be used individually or combined for a mixed microorganism population challenge
- Easily manipulated to deliver a wide variety of concentrations
- Available in concentrations ranging from 102 to 108 CFU per pellet
- ATCC™ 6633™†
Specifications
6633™* | |
1 | |
Lyophilized Pellet | |
10/Pk. |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures