Learn More
Fine Science Tools Micro Serrefines, 19mm
Supplier: Fine Science Tools 1805501
Product: Micro Serrefine - Straight/19mm/ Shape: Straight/ Jaw Dimensions: 10 x 2mm/ Clamping Pressure: Light/ Gram Pressure: 60g/ Alloy: Stainless Steel/ Length: 19mm/ Serrated: Yes/ Unit: Each