Learn More
Fine Science Tools Micro Needle Holder with Suture Cutter, 14 cm
Supplier: Fine Science Tools 1207514
Product: Micro Needle Holder with Suture Cutter/14cm/ Tip Shape: Curved/ Tip Width: 0.8mm/ Clamping Length: 5mm/ Scissors: Yes/ Lock: No/ Alloy: Stainless Steel/ Length: 14cm/ Serrated: No/ Unit: Each