Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mianserin hydrochloride, Tocris Bioscience™

Catalog Number 099750
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
21535-47-7
C18H21ClN2
300.83
MFCD00055072
YNPFMWCWRVTGKJ-UHFFFAOYNA-N
mianserin hydrochloride, mianserine hydrochloride, mianserin hcl, athymil, bolvidon, tolvon, tolvin, org gb 94, lerivon
68551
CHEBI:31843
hydrogen 5-methyl-2,5-diazatetracyclo[13.4.0.0²,⁷.0⁸,¹³]nonadeca-1(19),8,10,12,15,17-hexaene chloride
[H+].[Cl-].CN1CCN2C(C1)C1=CC=CC=C1CC1=CC=CC=C21
This item is not returnable. View return policy
21535-47-7
C18H21ClN2
300.83
MFCD00055072
YNPFMWCWRVTGKJ-UHFFFAOYNA-N
mianserin hydrochloride, mianserine hydrochloride, mianserin hcl, athymil, bolvidon, tolvon, tolvin, org gb 94, lerivon
68551
CHEBI:31843
hydrogen 5-methyl-2,5-diazatetracyclo[13.4.0.0²,⁷.0⁸,¹³]nonadeca-1(19),8,10,12,15,17-hexaene chloride
[H+].[Cl-].CN1CCN2C(C1)C1=CC=CC=C1CC1=CC=CC=C21

5-HT2 antagonist. Has moderate affinity for 5-HT6

Non-selective 5-HT2 receptor antagonist (Ki values are 0.0080 and 0.0081 μM for human 5-HT2A and 5-HT2C receptors expressed in HEK293 cells respectively). Has moderate affinity for 5-HT6. Antidepressant.
Quantity 50 mg
Formula Weight 300.83
Percent Purity >99%
Chemical Name or Material Mianserin hydrochloride
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit