Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO™ SevenCompact™ S220 pH Benchtop Meter

The S220 meter measures pH, ion concentration, and Redox/ORP. A simple and durable interface with only 2 buttons makes this meter very easy to operate, and results can be captured by a printer with a time and date stamp. Available as meter only or ready-to-use kits for specific applications.

Supplier:  METTLER TOLEDO™ 30096985

 View more versions of this product

Catalog No. 01-914-334

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

  • pH, Ion Concentration, and Redox/ORP benchtop meter that can display results in pH, mV, and Rel mV
  • Durable and simple to operate with two buttons: 'Read' and 'Cal'
  • IP54 Protection
  • Comes with electrode arm that moves straight up and down, bringing the electrode into position where it works best for your sample
  • Compatible with a wide range of electrodes for any application
  • Compatible with ISM electrodes that capture and save calibration data, which is then automatically recognized by the meter once connected
  • Meter connects to electrodes via BNC; S7 to BNC cables available
  • Results can be captured by a printer with a time and date stamp
  • Optional installation and qualification (IQ/OQ) package available through METTLER TOLEDO Service
TRUSTED_SUSTAINABILITY
Specifications

Specifications

±0.2 mV
S213 Kit: Micro Measurements
TFT
0.001/0.01/0.1
S220 pH/Ion meter, InLab Ultra-Micro-ISM electrode, InLab 741-ISM electrode, Multipin-BNC/RCA electrode cable, electrode holder, 2 buffer sachets for pH 4.01, 7, 9.21 and 10.01, 2 standards sachets 1413 uS/cm, pH Measurement Guide, protective cover, test certificate
pH/Ion Meter
±0.1°C
±0.002
Single channel
-2 to 20
-30.0°C to +130°C (MTC), -5.0°C to +130.0°C (ATC)
SC S220-Micro
0.1/1mV
MTC: -86° to +266°F, ATC: -41° to +266°F
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Accessories
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit