Learn More
Mettler Toledo™ MA130 Portable pH/Ion Meter
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
01-920-10
|
Mettler Toledo™
MA130BE |
Portable PH/ion Meter W/electrode | Portable PH/ion Meter W/electrode |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||
01-920-12
|
Mettler Toledo™
MA130FK |
-2 to +19.999 pH range | -2 to +19.999 pH range | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
Rugged and durable for use in both lab and field environments—so reliable, a battery change every 500 hours is the only internal maintenance it'll ever need. Ultrasonically welded casings are fully waterproofed to IP67 specifications—when used with special booted electrodes, the meter is dustproof and watertight to a depth of 1m for 30 minutes.
Measures pH (range -2.000 to +19.999) and ion concentration (range 0.000 to 19,999) with three-decimal-place precision. Can be used with most ISEs on the market. Also offers NTC and PT1000 temperature measurement, 0- to 9-hour last calibration reminder, adjustable slope and offset limits, and auto-increment sample ID.
Order Info
Packaging Options. Three packages are available; all packages include pH meter, batteries, instructions and an instrument sealing kit. The B (basic) package offers the meter without an electrode. The BE package includes an InLab™ 413 combination pH electrode with integrated temperature sensor and pH buffer sachets. Field Kits (FK) include the InLab combination electrode, pH buffer sachets, and a carrying case.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.