Learn More
Metrohm USA Metrosep A Trap 1 - 100/4.0
Supplier: Metrohm USA 61014000
High-capacity anion column, which is used to purify the eluent flow. Even chemicals of the highest quality level, e.g. "Ultrapur" or "puriss", can still contain minimal anion contaminations. These are reliably held back by the Metrosep A Trap 1 - 100/4.0. This column is primarily used with gradient applications.