Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxsalen, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog Number 18604404
Click to view available options
Quantity:
1 kg
Packaging:
Poly Pail

M1147, 298-81-7, C12H8O4

Spectrum™ Chemical Methoxsalen, USP is used in combination with UVA light therapy to treat severe psoriasis. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS 298-81-7
CAS Max % 100%
Ignition Residue 0.1%
Melting Range 143-148°C
Packaging Poly Pail
Molecular Formula C12H8O4
MDL Number MFCD00005009
Quantity 1 kg
UN Number UN1759
InChI Key QXKHYNVANLEOEG-UHFFFAOYSA-N
SMILES COC1=C2OC=CC2=CC2=C1OC(=O)C=C2
IUPAC Name 9-methoxy-7H-furo[3,2-g]chromen-7-one
Molecular Weight (g/mol) 216.19
Percent Purity 98 to 102%
Grade USP
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit