Learn More
MilliporeSigma™ Supelco™ Metal Packed GC Column (General Configuration), Phase: 30 Krytox 143AC, Matrix 60/80 Chromosorb P NAW Support
An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column is of a general configuration, and can be carefully bent to fit most instruments.
$614.26 - $1079.21
Specifications
Diameter (Metric) Inner 1.7 mm
For Use With (Application) Gas Chromatography (GC)
Material Copper Column
Packing Material Krytox 143 AC, Matrix 60/80 Chromosorb P NAW Support
Type Packed GC
View More Specs
Products 2
Catalog Number Mfr. No. Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Price Quantity & Availability  
11-100-5369
View Documents
MilliporeSigma™ Supelco™
13277U
12.2 m
Each for $1,079.21
Add to cart
11-100-8603
View Documents
MilliporeSigma™ Supelco™
13274U
6.1 m
Each for $614.26
Add to cart
Description
Description
Packed columns made up of stainless steel are basically employed when higher carrier gas flow rates are employed as they can sustain elevated pressure. Stainless steel columns are usually washed with dilute hydrochloric acid, then with water followed by methanol, acetone, methylene dichloride and n-hexane in order to remove any corrosion products and traces of lubricating agents
Specifications
Specifications
1.7 mm
Copper Column
Packed GC
Gas Chromatography (GC)
Krytox 143 AC, Matrix 60/80 Chromosorb P NAW Support