Learn More
MilliporeSigma™ Supelco™ Metal Packed GC Column (fits PerkinElmer), Phase: 0.1 SP-1000, Matrix 80/120 Carbopack C Support
An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column is of a general configuration, and can be carefully bent to fit most instruments.
Manufacturer:  MilliporeSigma™ Supelco™ 13736U
Catalog No. 11-101-0035
Add to cart
Description
Description
Specifications
Specifications
Gas Chromatography (GC)
Packed GC
Stainless Steel Column
2.1 mm
0.125 in.
1.8 m
SP-1000, Matrix 80/120 Carbopack C Support