Learn More
MilliporeSigma™ Supelco™ Metal Packed GC Column (fits Agilent, Configuration A), Phase: None, Matrix 80/100 Porapak S Support
An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column is of a general configuration, and can be carefully bent to fit most instruments.
Manufacturer:  MilliporeSigma™ Supelco™ 13161U
Catalog No. 11-101-0027
Add to cart
Description
Description
Specifications
Specifications
Gas Chromatography (GC)
0.125 in.
Stainless Steel Column
2.1 mm
Packed GC
1.8 m
Matrix 80/100 Porapak S Support