Learn More
MilliporeSigma™ Supelco™ Metal Packed GC Column (fits Agilent, Configuration A), Phase: None, Matrix 80/100 HayeSep D Support
An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column is of a general configuration, and can be carefully bent to fit most instruments.
Manufacturer:  MilliporeSigma™ Supelco™ 12921U
Catalog No. 11-100-9859
Add to cart
Description
Description
Specifications
Specifications
Gas Chromatography (GC)
Packed GC
Stainless Steel Column
0.75 mm
0.0625 in.
2 m
Matrix 80/100 HayeSep D Support