Learn More
MilliporeSigma™ Supelco™ Metal Packed GC Column (fits Agilent, Configuration A), Phase: 25 DC-200 (350 cstks), Matrix 80/100 Chromosorb P AW Support
An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column is of a general configuration, and can be carefully bent to fit most instruments.
Manufacturer:  MilliporeSigma™ Supelco™ 13039U
Catalog No. 11-100-8935
Add to cart
Description
Description
Specifications
Specifications
Gas Chromatography (GC)
0.125 in.
Stainless Steel Column
2.1 mm
Packed GC
4.6 m
DC-200 (350 cstks), Matrix 80/100 Chromosorb P AW Support