Learn More
MilliporeSigma™ Supelco™ Metal Packed GC Column (fits Agilent, Configuration A), Phase: 10 Carbowax 20M, Matrix 80/100 Chromosorb W AW Support
An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column is of a general configuration, and can be carefully bent to fit most instruments.
Manufacturer:  MilliporeSigma™ Supelco™ 12785U
Catalog No. 11-101-0032
Add to cart
Description
Description
Specifications
Specifications
Gas Chromatography (GC)
Packed GC
Stainless Steel Column
2.1 mm
0.125 in.
1.8 m
Carbowax 20 M, Matrix 80/100 Chromosorb W AW Support