Learn More
Lab-Aids™ Metabolism Experiment
Lab-Aids™ Metabolism Experiment uses a hands-on approach to introduce students to metabolic processes
Supplier: Lab-Aids™ 55
Description
- This kit introduces students to the study of metabolism by measuring the rate of oxygen consumption of small organisms (not included)
- Using Patented (U.S. Pat. No. 3,581,737) Lab-Aids Metabolism chambers, students observe organisms (not included) and draw correlations between size of an organism, the temperature of the environment, and oxygen consumption.
- They are also asked to calculate an organism's respiratory rate in cm3 of oxygen per kg per hour, measure total gas change in a closed environment and determine an organism's metabolic relationship to total gas in a closed environment.
- Includes two unbreakable chambers large enough for small animals and plants, one is used to measure the organism's oxygen consumption; the other serves as a control.
Specifications
U.S. Patent No. 3, 581, 737 | |
13 x 5 cm | |
Student Data Cards (10), Metabolism Chambers (2), Carbon Dioxide Absorbent (1 Bottle), Calibrated Capillary Tubes (1), Connecting Tube (1), Organism Holders (4) | |
∼50 min. |
6 to 12 | |
Metabolism Education | |
1 Kit | |
5 Students/Chamber |