Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meridian Bioscience™ Para-Pak™ Parasitology Transport Systems Reagents: Trichrome Blue Stain for Microsporidium
Fixatives, stain, and other reagents for use with Meridian Bioscience Para-Pak Parasitology Transport Systems
Supplier: Meridian Bioscience™ 400501
Specifications
Trichrome Stain | |
Dark Purple | |
5.5% Disodium 4,5-dihydroxy-3-phenylazonaphthalene-2,7-disulphonate, 2.87% Acetic Acid, 0.46% C.I. Acid Blue 22, 0.23% Tungsten Hydroxide Oxide Phosphate | |
1 Ea. | |
16.9 oz. | |
Stool |
CE | |
Liquid | |
Bottle | |
15°C to 30°C | |
500 mL |
Safety and Handling
ShelfLife : 18 Months
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction