Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meridian Bioscience™ illumigene™ C. difficile Test Kit
Molecular assay for the detection of C. difficiles in stool
Supplier: Meridian Bioscience™ 280050
Includes: Kits contain Sample Preparation Apparatus, test Device containing one dry reagent lyosphere in each of two chambers; a TEST Chamber with C. difficile specific primers a CONTROL Chamber with S. aureus specific primers necessary to complete 50 tests.
Description
illumigene kits were rebranded as Alethia. Product is identical only the name is different. No validation of Alethia is needed if currently using illumigene version. If discontinued illumigene kits are ordered the Alethia version will ship. New Number is 23029002
DNA Amplification Assay for the Detection of Cytotoxigenic C. difficile in Stool Specimens.
- Simple, flexible, cost-effective molecular assay for C. difficile detection.
- Allows addition of molecular assays without the cost of additional labor or capital expenditures.
- Specimen Type: Unformed stool
- Assay Format: Isothermal DNA amplification
- CE marked
- Sensitivity: 95.2%
- Specificity: 95.3%
Certifications
CE Marked
Specifications
CE Marked | |
Illumigene Clostridium difficile Test Kit | |
50 Tests |
C. difficile toxins A and B | |
Sample preparation apparatus, dry reagent Lyosphere, C. difficile specific primers, control chamber with S. aureus specific primers |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction