Learn More
Meridian Bioscience™ illumigene External Control Kit
for the detection of C. difficiles in stool
Supplier: Meridian Bioscience™ 279920
Description
illumigene kits were rebranded as Alethia. Product is identical only the name is different. No validation of Alethia is needed if currently using illumigene version. If discontinued illumigene kits are ordered the Alethia version will ship. New Number is 23029003
illumigene C. difficile External Control Reagents are supplied separately It is recommended that the reactivity of each new lot and each new shipment of illumigene C. difficile be verified on receipt and before use. External control tests should be performed thereafter in accordance with appropriate federal, state and local guidelines. The illumigene C. difficile test kit should not be used in patient testing if the external controls do not produce the correct results.
Specifications
CE Marked | |
Illumigene External Control Kit | |
18 Tests |
C. difficile | |
Positive and negative control reagent |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.