Learn More
Cole-Parmer Melting Point Calibration Kit
Supplier: Cole-Parmer 9996749
Accessories for your melting point apparatusEnsure the accuracy of your melting point apparatusComes with three WHO approved melting point standards—caffeine, vanillin, and sulphanilamide—with purity certificates.