Learn More
Chemglass Life Sciences Melting Point Tubes, Open Both Ends, 1.5mm-1.8mm O.D. x 0.20mm Wall, Vial of 100, 100mm Length
Supplier: Chemglass Life Sciences CG184001
Melting Point Tubes, Open Both Ends, 1.5mm-1.8mm O.D. x 0.20mm Wall, Vial of 100, 100mm Length. Tube is manufactured from borosilicate glass and packaged 100 tubes per vial. The Code -02 has a smaller I.D. to conform to U.S.P XXI requirements.PLEASE NOTE:SPECIFICATIONS FOR CG-1840-01 ARE O.D.