Learn More
Sigma Aldrich Fine Chemicals Biosciences Melittin from honey bee venom >=85% (HPLC) | 20449-79-0 | MFCD00076868 | 5MG
Supplier: Sigma Aldrich Fine Chemicals Biosciences M22725MG
Melittin from honey bee venom >=85% (HPLC) | Purity: >=85% (HPLC) | Mol Wt: 2846.46 | 20449-79-0 | MFCD00076868 | 5MG