Learn More
Medium Point Forceps
SCI_ED
Steel construction ensures years of service in every kind of dissecting work.
Supplier:  Medco Instruments 0114ACH
 View more versions of this product
Catalog No. S17323
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
Description
Description
  • Highly polished, chrome-plated steel forceps have file-cut, medium points for holding larger sections
  • Length: 3.98 in. (101mm)
Specifications
Specifications
Curved, Medium
Stainless Steel
Forceps, Dissecting
Curved
4 in.
101 mm
Each
Product Suggestions
Product Suggestions