Learn More
Zeflon International, Inc MEDIUM FLOW ROTAMETER 1-20LPM
Supplier: Zeflon International, Inc 195380
Medium Flow Rotameter Flowmeter, 1-20 LPM. Zefon Field Rotameters are a low cost way to measure airflow in air sampling applications. They feature a durable acrylic housing with direct reading scales printed on them. Available in three different flow ranges covering a range of 0.2 through 30 LPM. Operation and maintenance are very simple; only a few common sense precautions must be observed to assure long, trouble-free service.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.