Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Medi-First™ Antiseptic Wipes

Disinfects and cleans skin as first-aid antiseptic.

$4.13 - $10.71

Specifications

Product Type Antiseptic wipes
Composition 1:750 Benzalkonium chloride solution
Description Individual tamper-evident packets
For Use With (Application) Disinfects, cleans skin as first-aid antiseptic
Packaging Individual towelettes
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
19-090-836
View Documents
Medique
21433
100/Pk.
Each for $10.71
Case of 20 EA for $194.90
Edge
Add to Cart
 
19-090-835
View Documents
Medique
21471
20/Box
Each for $4.13
Case of 20 EA for $71.70
Add to Cart
 
Description

Description

  • Towelettes saturated with 1:750 Benzalkonium chloride solution
Specifications

Specifications

Antiseptic wipes
Individual tamper-evident packets
Individual towelettes
1:750 Benzalkonium chloride solution
Disinfects, cleans skin as first-aid antiseptic
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit