Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mechanix Wear™ M-Pact™ 2 Gloves - Black

Catalog Number p-313150
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

An accordion knuckle with EVA foam padding and Thermoplastic Rubber (TPR) finger guards protects against blunt force impact. D3O™ technology in the palm reduces impact and vibration from hand tools.

  • Thermoplastic Rubber (TPR) finger guards and metacarpal protection prevent impact injuries
  • Accordion knuckle guard with high density EVA foam padding
  • Armortex™ fingertips provide durable gripping power
  • D3O™ palm padding absorbs and dissipates high-impact energy in the palm
  • TrekDry™ material helps keep hands cool and comfortable
  • Extended Neoprene cuff provides added protection
  • Hook and Loop closure provides a secure fit
  • Machine washable
Cuff Material Neoprene
Color Black
Hand Specific
Material Thermoplastic Rubber
Product Type Mechanic's Gloves
Size Small
For Use With (Application) Construction, Heavy Equipment Operation, Maintenance & Repair, Manufacturing, Search & Rescue, and DIY Home Improvement
Glove Cuff Style Hook and Loop
Series M-Pact™ 2
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit