Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mechanix Wear™ FastFit™ Red Mechanics Gloves

Catalog Number p-304950
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

These high-dexterity, synthetic leather gloves are touchscreen compatible to help users stay connected while on the job. FastFit™ gloves offer ideal fit with TrekDry™ evaporative cooling technology to keep hands cool and comfortable.

  • TrekDry™ material is form-fitting to keep working hands cool and comfortable
  • High-dexterity synthetic leather infused with touchscreen technology
  • Stretch-elastic cuffs create a secure fit
  • Reinforced thumb and index finger provide additional durability
  • Anatomically designed two-piece palm design prevents material bunching
  • Pinched fingertip construction improves fingertip strength and durability
  • Machine washable
Back Material TrekDry™
Cuff Material Elastic
Coating Material Uncoated
Color Red
Hand Specific
Material Synthetic Leather
Product Type Mechanic's Gloves
Size Small
For Use With (Application) Maintenance Repairs, Equipment Operation, and DIY Home Improvement
Glove Cuff Style Open
Series FastFit™
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit