Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCR Safety Cut Pro™ 9676 Dyneema™ Cut-Resistant Gloves, Polyurethane Coating
Click to view available options
Size:
Small
Medium
Large
X-Large
Description
- 13-gauge salt and pepper colored DSM Certified Dyneema™ shell
- Cool and comfortable
- Cut-resistant, abrasion resistant, exceptional dexterity
- Gray polyurethane coated palm and fingertips
- Treated with ActiFresh™
Specifications
ANSI Abrasion Rating | ANSI 6 |
Coating Material | Polyurethane |
Color | Gray |
Cut Resistance Rating | ANSI A2 |
Material | Dyneema™ |
Product Type | Cut-resistant Gloves |
Size | Small |
For Use With (Application) | Abrasive applications, assembly, brick handling, construction, farming, general maintenance, landscaping, plumbing, recycling, shipping/receiving |
Glove Cuff Color | Yellow |
Glove Cuff Style | Knit |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction