Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCR Safety Eyeglass Cases
Click to view available options
Includes:
Belt Clip
Top flap, belt clip
Description
- Belt clip
- Vinyl material
- Color: Black
Specifications
Description | Eyeglass Case |
Color | Black |
Product Type | Eyewear case |
For Use With (Application) | Extends the life of protective eyewear |
Includes | Belt Clip |
Material | Vinyl |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction